Learn More
Molecular Probes™ protein G, horseradish peroxidase conjugate
protein G, horseradish peroxidase conjugate
Brand: Molecular Probes™ P21041
Code : 50
Additional Details : Weight : 0.00525kg
Description
The horseradish peroxidase (HRP) conjugate of protein G can be used to detect the Fc portion of various classes and subclasses of immunoglobulins from a variety of species. Protein G is a bacterial protein similar to protein A but with superior selectivity for mouse and human IgG.Specifications
HRP | |
Lyophilized Powder | |
Room Temperature |
1 mg | |
Store in freezer (-5 to -30°C). |
For Research Use Only. Not for use in diagnostic procedures.