missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Protein Kinase C-epsilon Peptide Substrate
For cellular and molecular biology applications
£67.12 - £101.61
Specifications
Color | White |
---|---|
Form | Lyophilized |
Molecular Weight (g/mol) | 2067.45 |
Storage Requirements | -30° to -10°C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White | |
Lyophilized | |
2067.45 | |
-30° to -10°C |
For Research Use Only