missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control

For cellular and molecular biology applications

Brand:  Thermo Scientific™ J66521.!D

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15430597

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Solid
843.98
0.5 mg
Leu-Ser-Glu-Thr-Lys-Pro-Ala-Val
-30°C to -10°C
A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.
Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
RUO - research use only
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only