missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protein LoBind™ Deepwell Plates
GREENER_CHOICEBIOPHARMA

Product Code. 10367133 Shop All Eppendorf Products
Click to view available options
Color:
White
White, Clear Bottom
Yellow
No. of Wells:
96
Volume (Metric) Well:
500 μL
1000 μL
2000 μL
Packaging:
10 Bags x 12 Plates
10 Bags x 8 Plates
5 Bags x 4 Plates
5 Bags x 8 Plates
Unit Size:
Pack of 120
Pack of 20
Pack of 40
Pack of 80

Product Code. 10367133

Brand: Eppendorf™ 0030508106

Only null left
Add to basket
Add to basket

Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.

  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.
TRUSTED_SUSTAINABILITY

Specifications

Certified Yes
Format Standard
Detection Method Quantitative protein analysis
Array 8 x 12
Autoclavable Autoclavable
Packaging 10 Bags x 12 Plates
Color White
Product Type Protein LoBind™ Plate
Material Polypropylene
Volume (Metric) Well 500 μL
No. of Wells 96
Sterility Non-sterile
Dnase Free Yes
For Use With (Application) Preparation or Storage of Protein, Peptide or Antibody Samples
Max. Heat Stability 121°C
Max. RCF 6000 x g
No. per Pack 12 Pk.
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 120 Pk.
RNase Free Yes
Well Color Colorless
Dimensions (L x W x H) 85.5 x 127.8 x 27.1 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.