Learn More
Eppendorf™ Protein LoBind™ Deepwell Plates
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
Brand: Eppendorf™ 0030504208
300.45 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 96
Additional Details : Weight : 2.25000kg
Description
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface ensure reliable sealing in plates.
Compliance
IVD, SBS/ANSI™
Specifications
IVD, SBS/ANSI | |
Standard | |
Quantitative protein analysis | |
Autoclavable | |
White, White | |
Polypropylene | |
96 | |
Non-sterile | |
Round Bottom (U-well) | |
Ideal for sample preparation or long-term storage of nucleic acids in forensic, microarrays, next generation sequencing applications | |
6000 x g | |
PCR Clean (Non-sterile) | |
Yes | |
-86°C to +100°C | |
85.5 x 127.8 x 44.1 mm | |
4.41 cm |
Yes | |
Without Lid | |
8 x 12 | |
20 Plates (5 Bags x 4 Plates) | |
Protein LoBind™ Plate | |
1000 μL | |
30 to 1,000 μL | |
Protein LoBind | |
Yes | |
121°C | |
20/Pk | |
20 Pk. | |
-122.8°F to 212°F | |
Colorless | |
12.78 cm | |
8.55 cm |