Learn More
Cytiva Whatman™ Protein Prep for ӒKTA™ Chromatography Systems
The Protein Prep syringe filter containing the regenerated cellulose membrane is the optimal choice for filtration prior to chromatography with ӒKTA™ systems.
£424.00 - £465.00
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | Price | Quantity & Availability | ||||
15851838
|
Cytiva
10463043 |
0.2 μm | 30 mm |
£465.00
Pack of 150 |
In Stock
Log in to see stock. |
||||
15861838
|
Cytiva
10463103 |
0.2 μm | 13 mm |
£424.00
Pack of 150 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
15871838
|
Cytiva
10463113 |
0.45 μm | 13 mm |
£424.00
Pack of 150 |
In Stock
Log in to see stock. |
||||
15841838
|
Cytiva
10463033 |
0.45 μm | 30 mm |
£465.00
Pack of 150 |
In Stock
Log in to see stock. |
||||
Description
Specifications
150 Pieces |