Learn More
Novus Biologicals™ Protein S/PROS1 Overexpression Lysate
Brand: Novus Biologicals™ NBL1-14812
Additional Details : Weight : 0.00970kg
Description
The Protein S/PROS1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC2273 and based on accession number NM_313. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens protein S (alpha) (PROS1), mRNA. | |
Native |
For Research Use Only