Learn More
Thermo Scientific™ Proteinase K, recombinant, PCR grade
Degrade proteins even in the presence of detergents with Proteinase K, a broad-range endolytic protease widely used for digestion of proteins in nucleic acid preparations.
£35.59 - £147.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10181030
|
Thermo Scientific™
EO0491 |
1 mL |
£35.59
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11501515
|
Thermo Scientific™
EO0492 |
5 x 1 mL |
£147.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proteinase K is an endolytic protease that cleaves peptide bonds at the carboxylic sides of aliphatic, aromatic, or hydrophobic amino acids. The Proteinase K is classified as a serine protease. The smallest peptide to be hydrolyzed by this enzyme is a tetrapeptide.
Highlights
- Ready-to-use solution
- Active in a wide range of reaction conditions
Specifications
Proteinase K | |
Liquid |
Safety and Handling
- Proteinase K
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : Proteinase
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.