Learn More
Abeba™ Protektor Line 2230 Shoes
Leather upper construction includes reflective material.
Brand: Abeba™ 2230/40
Additional Details : Weight : 1.22000kg
Description
- CE EN ISO 20345:2011 S3 SRC
- Smooth leather white
- Steel toe cap
- Penetration resistant steel midsole
- Slip resistance PU outsole sole
- Antistatic
- Leather lining
- Double velcro fastening
Specifications
CE EN ISO 20345:2011 S3 SRC | |
Leather | |
Slip-resistant | |
2230 |
White | |
40 | |
Polyurethane | |
Steel |