Learn More
Abeba™ Protektor Line 2230 Shoes
Leather upper construction includes reflective material.
Brand:  Abeba™ 2230/46
Additional Details : Weight : 1.22000kg
 View more versions of this product
Product Code. 15678027
  • £76.87 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather white
  • Steel toe cap
  • Penetration resistant steel midsole
  • Slip resistance PU outsole sole
  • Antistatic
  • Leather lining
  • Double velcro fastening
Specifications
Specifications
CE EN ISO 20345:2011 S3 SRC
Leather
Slip-resistant
2230
White
46
Polyurethane
Steel