Learn More
Novex™ ProtoArray™ Blocking Buffer Kit
ProtoArray™ Blocking Buffer Kit
Brand:  Novex™ PA055
Code : NEW
Additional Details : Weight : 0.14000kg
Product Code. 10033344
  • £146.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
Description
Description
The Blocking Buffer Kit consists of reagents sufficient for preparing Blocking Buffer and Wash Buffer to process 10 ProtoArray™ Human Protein Microarrays for ProtoArray™ protein-protein interaction (PPI), immune response biomarker profiling (IRBP), and antibody specificity profiling (ASP) applications. The kit contains one bottle each of Blocking Buffer and 10X Synthetic Block, and one tube of dithiothreitol (DTT).
Specifications
Specifications
ProtoArray™ Blocking Buffer Kit, 1 Kit, –20°C Storage Temperature, For Applications Pharma and Biopharma, Protein Assays and Analysis, Target And Lead Identification And Validation
1 Kit
-20°C
Protein Array Blocking Buffer
Protein Assay and Analysis