missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocadherin alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£274.29 - £392.00
Specifications
Antigen | Protocadherin alpha 1 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18400811
|
Novus Biologicals
NBP1-86297-25ul |
25 μL |
£274.29
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18283636
|
Bio-Techne
NBP1-86297 |
0.1 mL |
£392.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protocadherin alpha 1 Polyclonal specifically detects Protocadherin alpha 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Protocadherin alpha 1 | |
Polyclonal | |
Rabbit | |
Human | |
KIAA0345-like 13, PCDH-alpha-1, PCDH-ALPHA1, protocadherin alpha 1, protocadherin alpha-1 | |
PCDHA1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
56147 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LSPSLNTSERNEQPEANLDLSGNVSPTFEF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only