missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-3, 12-20 μm Bulk Resin

Reversed-phase column optimized for separation of macromolecules such as DNA, RNA oligos, proteins, peptides and proteomics. Hamilton’s PRP-3 is a polymeric reversed-phase HPLC column designed for the purification and isolation of proteins and peptides with very good recovery (> 90%).

Brand:  Hamilton™ 79702

Code : NEW

Additional Details : Weight : 0.17000kg

Product Code. 11567874

  • £94.50 / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

Specification Sheet

Separation ModeReversed-Phase
Stationary PhasePRP-3
FormatBulk Resin
Particle Size12-20 μm
Packing Material TypePSDVB
Pore Size300 Å
Max Pressure5,000 psi
RestorationRun a gradient of 100% water to 100% acetonitrile (Repeat three times)
UOMper gram
Specifications

Specifications

12 to 20 μm
Bulk Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers