missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ PS-M3D Variable Speed / Angle, Multi-function 3D Rotator

PS-M3D Operates continuously or can be set for timed operation. Grant Bio™ PS-M3D Variable Speed / Angle, Multi-function 3D Rotator with individual phases may be linked together for absolute operational flexibility.

Brand:  Grant Instruments™ PS-M3D

Product Code. 12985501

  • £977.00 / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vibration - short wave motion, best for mixing small particles
  • 3D rotation speed: 1 to 100 rpm
  • Reciprocal 3D rotation: 1 to 360° turning angle
  • Vibration: 1 to 6° turning angle, programmable in a burst of 1 to 5 seconds
  • Fully programmable sequence of functions Loads up to 1.0 kg
  • Very easy to operate, with simple set-up of multi-segment programs via push buttons and the 2-line LCD status display
  • Compact, with a low profile and small footprint, and extremely quiet in operation - fits neatly and unobtrusively into the workspace
  • Smooth, non-slip mat supplied as standard - prevents vessels from slipping
  • Continuous or timed operation
  • Ambient operating temperature range of 4 to 40°C allows use in cold room or incubator
TRUSTED_SUSTAINABILITY
Specifications

Specifications

22 mm
1 kg
1 to 100 rpm
4°C to 40°C
0.38 A
235 mm
140 mm
235 mm
2 x 16 character LCD
7°C
  • 0 to 360°, 30° increments (reciprocal mode)
  • 0 to 5°,  1° increments (
120/230 V
4.6 W
50/60 Hz
215 x 215 mm
235 mm
PS-M3D
  • 0 to 250sec
  • 0 to 5 (vibration mode)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Grant Instruments™ PS-M3D Variable Speed / Angle, Multi-function 3D Rotator >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.