missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ PS MicroKos™ Filter Modules
Spectrum Labs™ PS MicroKos™ Filter Modules are single-use, hollow fiber filters designed for tangential flow filtration.
£123.00
Specifications
Material | Polysulfone |
---|---|
Sterility | Non-sterile |
Includes | Inlet/Outlet Ports: Male Luer-Lok, Side Port: Female Luer-Lok |
Type | Hollow Fiber Filtration Module |
Diameter (Metric) | Housing OD: 4.6 mm |
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | MWCO | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | MWCO | Price | Quantity & Availability | |||
12407091
|
Spectrum Labs™
C02-S500-05-N |
Fiber ID: 0.5mm, Housing ID: 4.6mm, Inlet/Retentate ID: 3mm, Side Port ID: 4.3mm | Total Length: 23cm, Effective Length: 20cm | 500 kDa |
£123.00
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||
Description
- Designed for tangential flow filtration of small volumes ranging from 1mL to 100mL
- Single-use MicroKros™ hollow fiber filters are the first efficient and practical tangential flow filtration device for characterizing a process for scale-up or filtration of small volumes in a laboratory setting.
- MicroKros™ are available with MLL x FLL connections and 1 Effective Length
Specifications
Polysulfone | |
Inlet/Outlet Ports: Male Luer-Lok, Side Port: Female Luer-Lok | |
Housing OD: 4.6 mm |
Non-sterile | |
Hollow Fiber Filtration Module |