missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSB 0777 ammonium salt, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent adenosine A2A receptor full agonist (Ki = 44.4 nM for rat brain striatal A2A receptors). Exhibits subtype selectivity for A2A receptors over A1, A2B and A3 receptors (Ki ≥ 10 000 nM for rat A1, human A2B and human A3 receptors). Potentiates acetylcholine-induced contractions in ex vivo rat ileum/jejunum segments.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 500.55 |
Percent Purity | >98% |
Chemical Name or Material | PSB 0777 ammonium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'