missing translation for 'onlineSavingsMsg'
Learn More

PSB 0777 ammonium salt, Tocris Bioscience™

Product Code. 30001492
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent adenosine A2A agonist

Potent adenosine A2A receptor full agonist (Ki = 44.4 nM for rat brain striatal A2A receptors). Exhibits subtype selectivity for A2A receptors over A1, A2B and A3 receptors (Ki ≥ 10 000 nM for rat A1, human A2B and human A3 receptors). Potentiates acetylcholine-induced contractions in ex vivo rat ileum/jejunum segments.

Chemical Identifiers

CAS 2122196-16-9
Molecular Formula C18H24N6O7S2
Molecular Weight (g/mol) 500.55
InChI Key AYQGFWKUGKSGCZ-UHFFFAOYNA-N
Synonym psb 0777 ammonium salt, 4-2-6-amino-9-b-d-ribofuranosyl-9h-purin-2-yl thio ethyl benzenesulfonic acid ammonium salt
PubChem CID 90488932

Specifications

Quantity 50 mg
Formula Weight 500.55
Percent Purity >98%
Chemical Name or Material PSB 0777 ammonium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'