missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSB 0788, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective adenosine A2B receptor antagonist (IC50 = 3.64 nM at human A2B receptors). Displays > 100-fold selectivity over hA1, hA2A and hA3 receptors.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 543.04 |
Percent Purity | >98% |
Chemical Name or Material | PSB 0788 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'