Learn More
PSI-6206 13C,d3, MedChemExpress
MedChemExpress PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
POA - £2162.27
Specifications
Chemical Name or Material | PSI-6206 13C,d3 |
---|---|
CAS | 1256490-42-2 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C9 13CH10D3FN2O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16451489
|
Medchem Express
HY-15236S |
1 mg |
£504.57
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16436388
|
Medchem Express
HY-15236S |
10 mg |
POA
10mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16418977
|
Medchem Express
HY-15236S |
5 mg |
£2162.27
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.Specifications
PSI-6206 13C,d3 | |
Light Yellow | |
C9 13CH10D3FN2O5 | |
OC[C@@H]1[C@H]([C@@]([13C]([2H])([2H])[2H])(F)[C@H](N2C(NC(C=C2)=O)=O)O1)O | |
264.23 | |
Research |
1256490-42-2 | |
Research | |
RO-2433 13C,d3 GS-331007 13C,d3 Sofosbuvir metabolite GS-331007 13C,d3 | |
264.23 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.