missing translation for 'onlineSavingsMsg'
Learn More

PSI-6206, MedChemExpress

Product Code. p-200099502
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16432497

Brand: Medchem Express HY15236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PSI-6206 (RO 2433) is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.

PSI-6206 (RO 2433) is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.

Specifications

Chemical Name or Material PSI-6206
CAS 863329-66-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H13FN2O5
Quantity 50 mg
Synonym RO 2433 GS-331007
Solubility Information DMSO : 100 mg/mL (384.29 mM; Need ultrasonic)
SMILES O=C(NC1=O)C=CN1[C@H]2[C@](F)([C@@H]([C@H](O2)CO)O)C
Molecular Weight (g/mol) 260.22
Formula Weight 260.22
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.