missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14748-1-AP-20UL
Weitere Details : Weight : 0.00010kg
Beschreibung
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the non-ATPase subunits of the 19S regulator lid. In addition to participation in proteasome function, this subunit may also participate in the TNF signalling pathway since it interacts with the tumor necrosis factor type 1 receptor. A pseudogene has been identified on chromosome 1.Spezifikation
PSMD2 | |
Polyclonal | |
Unconjugated | |
PSMD2 | |
26S proteasome subunit p97, P97, Protein 55.11, PSMD2, Rpn1, S2, TRAP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21762, 287984, 5708 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13200, Q4FZT9, Q8VDM4 | |
Psmd2 | |
PSMD2 Fusion Protein Ag6484 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |