Learn More
BINDER™ Pt 100 Temperature Sensor
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers KT 53
Unit Size:
Each
Description
- Flexible
- Additional sensor
- Interior
Specifications
Specifications
| Description | Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display |
| For Use With (Equipment) | BINDER Chambers KT 53 |
| Material | Plastic |
| Type | Temperature Sensor |