missing translation for 'onlineSavingsMsg'
Learn More

Kinematica™ 12 mm M-Design Aggregate for Polytron™ for PT 1200 E, PT 1300 D and PT 2500E Homogenizers

Cut everything with merciless protruding blades, and complete the dispersion process in one pass without changing your aggregate. Kinematica™ PT-DA M Aggregates are designed for use with POLYTRON Batch Dispensers: PT 1200 E, PT 1300 D, PT 2500 E and PT 4000.

Brand:  Kinematica™ 11030266

Product Code. 11960439

  • £1905.00 / Each
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rotor with integrated blades
  • Standard rotor/stator design 
  • Two rows of teeth and blades
  • All aggregates are delivered with all necessary assembly / disassembly tools
  • Cuts sample outside the generator
  • Processes of large samples or particles
  • Rotor with blades facilitates processing of larger cell tissue samples or particles
  • Dispersing and mixing of particles with a size of up to 25mm
  • Sample preparation for the extraction and solving of organic material
  • Dispersing of fibrous, stringy and hard samples (ex. meat)
Specifications

Specifications

Aggregate
12 mm
Rotor with 2 rows of teeth and integrated blades
POLYTRON™ Batch Dispensers: PT 1200 E, PT 1300 D, PT 2500 E, PT 4000
2 mL to 30 L
M-design
Sample preparation for the extraction and solving of organic material, processing large samples or particles and dispersing of fibrous, stringy and hard samples (e.g., meat)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers