missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH PTFE Adapters
£413.00 - £499.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10641414
|
RETSCH
22.008.0005 |
For 5 Reaction Vials 1.5 and 2 mL |
£499.00
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
10677313
|
RETSCH
22.008.0006 |
For 10 Reaction Vials 0.2 mL |
£413.00
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
Specifications
Adapter | |
PTFE |