Learn More
Fisherbrand™ PTFE Anchor Propeller Stirrer Shaft
Use with shaft guides for mixing high viscosity substances at slower speeds. The PTFE anchor stir shafts have a stainless-steel core.
£36.35 - £118.00
Specifications
For Use With (Equipment) | Universal Stirrer Guide |
---|---|
For Use With (Application) | Mixing |
Quantity | 1 Each |
Temperature (Metric) | -200°C to +280°C |
Product Code | Brand | Diameter (Metric) Shaft | Length (Metric) | Diameter (Metric) Fan | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Shaft | Length (Metric) | Diameter (Metric) Fan | Price | Quantity & Availability | |||
10446101
|
Fisherbrand™
|
10 mm | 650 mm (EX) | 140 mm |
£118.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10418791
|
Fisherbrand™
|
9.5 mm | 750 mm (EX) | 140 mm |
£63.80
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10456101
|
Fisherbrand™
|
12 mm | 650 mm (EX) | 140 mm |
£83.80
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
10339880
|
Fisherbrand™
|
9.5 mm | 650 mm (EX) | 140 mm |
£59.20
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
10197651
|
Fisherbrand™
|
12 mm | 1000 mm (EX) | 140 mm |
£92.85
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
10085921
|
Fisherbrand™
|
10 mm | 750 mm (EX) | 140 mm |
£80.55
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10341592
|
Fisherbrand™
|
12 mm | 750 mm (EX) | 140 mm |
£84.90
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
10526712
|
Fisherbrand™
|
9.5 mm | 550 mm | 140 mm |
£64.65
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
10470751
|
Fisherbrand™
|
9.5 mm | 650 mm | 140 mm |
£61.55
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
10646681
|
Fisherbrand™
|
9.5 mm | 550 mm | 100 mm |
£55.05
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
10269891
|
Fisherbrand™
|
8 mm | 500 mm | 80 mm |
£51.50
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10516712
|
Fisherbrand™
|
9.5 mm | 650 mm | 100 mm |
£59.50
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10780333
|
Fisherbrand™
|
10 mm | 550 mm | 100 mm |
£73.50
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10115590
|
Fisherbrand™
|
6 mm | 500 mm | 80 mm |
£52.63
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10496861
|
Fisherbrand™
|
8 mm | 400 mm | 80 mm |
£56.10
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10248020
|
Fisherbrand™
|
6 mm | 400 mm | 80 mm |
£37.98
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10629542
|
Fisherbrand™
|
8 mm | 300 mm | 80 mm |
£45.60
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
10637501
|
Fisherbrand™
|
6 mm | 300 mm | 80 mm |
£36.35
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
Description
PTFE shaft stirrers have a unique, fully molded construction featuring a rotor molded to a fully PTFE-encapsulated steel reinforced drive shaft.- Chemically resistant, inert, and leak-free
- Easy to clean
- No cross contamination
- Shatterproof
- Non-scratch
- Use in temperatures up to approximately 280°C
- Exposed shaft models (EX) are produced with a stainless steel core with an exposed end for more rigid clamping to the chuck drive
- For mixing high viscosity substances at slower speeds
Specifications
Universal Stirrer Guide | |
Mixing | |
1 Each | |
-200°C to +280°C |