missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTFE Beaker Cover-Watch Glass
Click to view available options
Description:
Watch glass; Fits 1000mL Beakers
Watch glass; Fits 100mL Beakers
Watch glass; Fits 2000mL Beakers
Watch glass; Fits 250mL Beakers
Watch glass; Fits 25mL Beakers
Watch glass; Fits 5-10mL Beakers
Watch glass; Fits 500mL Beakers
Watch glass; Fits 50mL Beakers
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Available in a range of diameters
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Description | Watch glass; Fits 2000mL Beakers |
Material | PTFE |
Type | Beaker |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'