missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Beaker with Spout

Catalog Number 16236511
Click to view available options
Capacity (Metric):
1 mL
5 mL
10 mL
25 mL
50 mL
100 mL
150 mL
250 mL
400 mL
500 mL
600 mL
1000 mL
2000 mL
5000 mL

Includes a machined base for good heat transfer. The PTFE beaker includes a molded-in spout for easy pouring.

Non-contaminating and inert to highly corrosive reagents.
  • Non-wettable, lightweight, and heat resistant up to 280°C
  • Machined base for good heat transfer
  • Easy to clean
  • Molded-in spout for easy pouring
  • Isostatically molded from pure PTFE
Color White
Diameter (Metric) Outer 13mm
Diameter (Metric)Top 13mm
Capacity (Metric) 1 mL
Material PTFE
Height (Metric) 18mm
Temperature (Metric) Max. Operating 280°C
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'