missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cowie Technology™ PTFE Screw-top Bottle
24.46 GBP valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Use promo code "21638" to get your promotional price.
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
150 mL
500 mL
1000 mL
2000 mL
Diameter (Metric):
22 mm
26 mm
33 mm
43 mm
52 mm
60 mm
80 mm
100 mm
120 mm
Diameter (Metric) Inner Neck:
10mm
12mm
19mm
25mm
35mm
52mm
57mm
60mm
Description
- Isostatically molded in pure PTFE and with super smooth internal finish, FDA approved
- Insoluble and non-contaminating
- Exceptional thermal resistance can be used from -200°C to +280°C
- Hand grips for ease of manipulation
- Do not use, when sealed, for chemical reactions or digestions, loosen lid when thermal sterilizing
Specifications
Closure Type | Screw Cap |
Diameter (Metric) | 52 mm |
Product Type | Container |
Capacity (Metric) | 100 mL |
Color | White |
Diameter (Metric) Inner Neck | 35mm |
For Use With (Application) | Storage of hazardous chemicals |
Height (Metric) With Closure | 88mm |
Material | PTFE |
Max. Temperature (Metric) | 280°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'