missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ PTFE Bottles Product Code.: 15466185

Thermo Scientific™ PTFE Bottles

Product Code. 15466185
250 mL, 115 mm
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1 L
2000 mL
Height (Metric):
26 mm
61 mm
76 mm
80 mm
88 mm
100 mm
115 mm
150 mm
185 mm
240 mm
This item is not returnable. View return policy

Product Code. 15466185

Brand: Thermo Scientific™ 038113.KT

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with aggressive or valuable reagents

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Inert and leak-free
  • Flexible and with PTFE cap
TRUSTED_SUSTAINABILITY

Specifications

Capacity 250 mL
Capacity (Metric) 250 mL
Material PTFE
Content And Storage Ambient temperatures
Height (Metric) 115 mm
Product Type Reagent Bottle
Diameter (Metric) 67 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ PTFE Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.