Learn More
BRAND™ PTFE Coated Steel Core Stirring Rods
Unique high-density PTFE coated polyethylene stirring rods with a steel core with rounds ends.
Brand:  BRAND™ 136325
 View more versions of this product
Product Code. 10029880
  • £23.20 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PTFE coated steel core stirring rods with round end
  • High-density polyethylene (with steel core)
  • Can be bent as required
  • The shape is permanently retained
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE Coated Steel Core
6 mm
300 mm
1