Learn More
BRAND™ PTFE Coated Steel Core Stirring Rods
Unique high-density PTFE coated polyethylene stirring rods with a steel core with rounds ends.
Brand: BRAND™ 136320
14.25 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- PTFE coated steel core stirring rods with round end
- High-density polyethylene (with steel core)
- Can be bent as required
- The shape is permanently retained
Specifications
PTFE Coated Steel Core | |
6 mm | |
250 mm | |
1 |