Learn More
Fisherbrand™ PTFE Conical Joint Clamps
Use as a safe solution to clamping glass joints. The PTFE conical joint clamps include an encapsulated steel spring.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 16266031
  • £40.50 / Pack of 10
Estimated Shipment: 21-10-2024
to see stock.
Add to basket
Description
Description
PTFE joint clamps are chemically inert and chemically resistant. Temperature resistant, maintaining strength up to 280°C (480°F). Manufactured from pure PTFE, with totally encapsulated steel springs. They can be adjusted by hand and fit standard glass joints.
Specifications
Specifications
Conical
PTFE Joint Clamp
Standard glass joints, size 12
2/Pk.
PTFE-encapsulated Steel Spring
White