missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
Fisherbrand™ PTFE Crucible | |
1 | |
60 mm |
PTFE | |
100 mL | |
45 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ PTFE Crucible > ml: 100; Height: 45mm; OD: 60mm
Spot an opportunity for improvement?Share a Content Correction