Learn More
Bohlender™ PTFE Cylindrical Magnetic Stirring Bar
Brand: Bohlender™ C350-23
This item is not returnable.
View return policy
Specifications
FDA Certified | |
Alnico 5 magnetic core | |
6 mm | |
PTFE | |
Cylinder |
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics | |
PTFE | |
35 mm | |
1/Pk. | |
Magnetic Stir Bar |