missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ PTFE Diagnostic Slides

These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use

Brand:  Epredia™ X2XER203B

109.20 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.


 View more versions of this product

Product Code. 10632391

  • £162.00 / Pack of 100
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Binds and keeps liquids applied to the well area
  • PTFE Construction
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
Specifications

Specifications

25 mm
PTFE
3
90°, ground edge
100/Pk.
75 mm
Diagnostic Slide
Black
14 mm Well
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers