missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Funnel

Includes a super smooth non-stick internal surface. the PTFE funnel is chemically resistant and inert.

Brand:  Fisherbrand™ 039.150

 View more versions of this product

Product Code. 10620463

  • £192.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Chemically inert with super smooth, non-stick internal finish.
  • Heavy wall construction
  • Use up to 280°C
  • Isostatically molded in pure PTFE
Specifications

Specifications

150mm
PTFE
60°
PTFE Funnels
84mm
16mm
220 mm
Autoclavable
Addition of liquids to vessels
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.