Learn More
Kartell™ Lining for Glass Tapers
Constructed of PTFE for high temperature and chemical resistance. Kartell™ Lining for Glass Tapers are specially designed to prevent machined glass joints from binding.
£27.90 - £40.95
Specifications
Certifications/Compliance DIN 12242
Material PTFE
For Use With Glass Joint
For Use With (Application) Prevent glass joints from binding
Packaging Quantity 10/Pk.
View More Specs
Products 5
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
10554851
View Documents
Kartell™
0103800
14/23 Cone Size
£27.90
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
10234851
View Documents
Kartell™
0103900
19/26 Cone Size
£28.35
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
10717741
View Documents
Kartell™
0104100
29/32 Cone Size
£35.50
Pack of 10
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
10707741
View Documents
Kartell™
0104000
24/29 Cone Size
£32.80
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
15845497
View Documents
Kartell™
0104300
45/40 Cone Size
£40.95
Pack of 10
Estimated Shipment: 15-08-2025
to see stock.
Add to basket
Description
Description
  • Designed to prevent machined glass joints from binding
  • Conforms to DIN 12242
  • Suitable for foodstuff
Specifications
Specifications
DIN 12242
Glass Joint
10/Pk.
PTFE
Prevent glass joints from binding
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Kartell™ Lining for Glass Tapers