Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, bent
For safe connection of hoses.
Brand: DWK Life Sciences 286230605
Code : NEW
Additional Details : Weight : 0.07000kg
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
- Hose olives enable the connection of hoses to laboratory apparatuses
Specifications
10/Pk. | |
Bent | |
9 mm | |
GL 14 |