Learn More
Welch™ PTFE Inline Filter
Economical in-line particulate filter maintains the cleanliness and proper operation of your vacuum pump. Welch™ PTFE Inline Filter protects your vacuum pump from fine particulates and aerosols to 0.2 microns.
Brand: Welch™
Code : 04
Additional Details : Weight : 0.01800kg
Description
- Protects vacuum pump from particulates
- 0.2 micron pore size
- PTFE filter
- Polypropylene housing
Specifications
Autoclavable | |
0.2 μm | |
PP | |
Inline Filter, DN 8 |
PTFE | |
Particulate filtration, vacuum pump | |
1 Each |