missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Containers
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
120 mL
240 mL
360 mL
480 mL
1000 mL
2200 mL
Specifications
Specifications
Capacity (Metric) | 240 mL |
Capacity (English) | 8 oz. |
Color | White |
Diameter (Metric) Outer Neck | 63mm |
Diameter (Metric) Inner Neck | 60mm |
For Use With (Application) | Storage and Transport of Hazardous Chemicals |
Includes | Closure |
Height (Metric) With Closure | 97mm |
Material | PTFE |
Max. Temperature (Metric) | 280°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'