Learn More
Quickfit™ PTFE Joint Clips with Conical Joints
Highly resistant to chemical attack
Brand: Quickfit GC34
This item is not returnable.
View return policy
Description
Highly resistant to chemical attack
- Used for retaining glassware having Quickfit™ conical joints
- Manufactured from PTFE with encapsulated steel spring to provide firm grip
- Clips are numbered with joint size for easy identification
- Can be used at temperatures up to 250°C
Specifications
Joint Clip | |
Encapsulated Steel spring | |
34/35 |
PTFE | |
Conical Joints | |
250°C |