Learn More
DWK Life Sciences PTFE Key with Thread, with three part retaining device
For use with polished DURAN™ disposable taps.
Brand:  DWK Life Sciences 286220407
Code : Z4
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 12858041
  • £358.00 / Pack of 50
Estimated Shipment: 09-09-2024
to see stock.
Add to basket
Description
Description
Disposable PTFE valve plugs in combination with the polished DURAN™ disposable valve represent a grease-free alternative. The 3-part tap safety device is already included in the scope of delivery.
  • Made of PTFE with thread and 3-part valve locking device
  • For use with polished DURAN™ one-way taps
  • Grease-free working
Specifications
Specifications
2.5 mm
No
50/Pk.
PTFE
14.5 NS
For use with polished DURAN™ one-way taps