missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ PTFE Keys
£15.76 - £18.93
Product Code | Brand | Diameter (Metric) | Fits Tubing ID | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Fits Tubing ID | Price | Quantity & Availability | |||||
10135682
|
Lenz Laborglasinstrumente™
02171804 |
4mm | 18.8 |
£18.93
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10145682
|
Lenz Laborglasinstrumente™
02171402 |
2.5mm | 14.5 |
£15.76
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10160613
|
Lenz Laborglasinstrumente™
02171404 |
4mm | 14.5 |
£16.85
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
Specifications
Stopcock Key | |
PTFE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ PTFE Keys