Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Brand:  Fisherbrand™ 001.807
 View more versions of this product
Product Code. 11888952
  • £17.45 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
Specifications
2 mm
7 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ PTFE Micro Magnetic Stir Bar > Dimensions (Dia. x L): 2 dia.x 7mm