Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Brand:  Fisherbrand™ 001.830
 View more versions of this product
Product Code. 16245851
  • £20.30 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
30 mm
Cylinder
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
3 mm
5/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ PTFE Micro Magnetic Stir Bar > Dimensions (Dia. x L): 3 dia.x 30mmL