Learn More
Fisherbrand™ Octagonal Magnetic Stir Bar Assortment with Retainer
Provides greater surface area and added turbulence when compared to smooth cylindrical bars. Fisherbrand™ Octagonal Magnetic Stir Bar Assortments with Restrainers have an integral pivot ring on each bar to aid in reducing friction and chattering.
Brand:  Fisherbrand™ F37158-0010
Product Code. 11893210
  • £79.05 / Each
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Alnico magnets encapsulated in FDA grade low friction coating
  • Restrainer fits comfortably in one hand
  • Eliminates splashing hazards when pouring
Order Info
DimensionsLxW
Specifications
Specifications
ISO 9001:2008
Integral Pivot Rings, Spinbar Restrainer
PTFE
Octagon
12.7 x 3.175, 12.7 x 7.92, 38.1 x 7.92, 50.8 x 7.92mm
PTFE
1/Ea.
Magnetic Stir Bar, Micro