Learn More
Thermo Scientific™ PTFE One-Piece HPLC Column Connector
Ensure leak-free column connections and optimize liquid chromatography performance with the inert and biocompatible Thermo Scientific™ PTFE One-Piece HPLC Column Connector.
Brand: Thermo Scientific™ 60170-370
95.41 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 16
Additional Details : Weight : 0.00090kg
Description
Minimize HPLC system dead volume and ensure optimal separation efficiencies with the Thermo Scientific™ PTFE One-Piece HPLC Column Connector. Ideal for coupling analytical and guard columns with 10–32 threads, it provides fingertight, leak-free connections. The material is inert and biocompatible for a wide range of applications.
Specifications
Coupler | |
One-piece column coupler | |
PEEK | |
Analytical and Guard Columns with 10 to 32 Threads |