missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Platinum Resistance Probe with 1M Cable
Inert to virtually all chemical reagents. Fisherbrand™ PTFE Thermocouple Type K Probe combine the outstanding properties of PTFE with the accuracy and reliability of platinum resistance and thermocouple thermometry.
£66.30 - £117.00
Specifications
Type | Probe, PTFE Platinum Resistance |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Material | PTFE with Stainless Steel Core |
Sensor Types | PRT100 Class A. TCR = 3850 ppm/degree celsius |
For Use With (Application) | Measuring temperature |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11752464
|
Fisherbrand™
500.500.1 |
500 mm |
£89.55
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11762464
|
Fisherbrand™
500.700.1 |
700 mm |
£117.00
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11722464
|
Fisherbrand™
500.200.1 |
200 mm |
£72.40
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11712464
|
Fisherbrand™
500.100.1 |
100 mm |
£66.30
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11742464
|
Fisherbrand™
500.400.1 |
400 mm |
£80.90
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11732464
|
Fisherbrand™
500.300.1 |
300 mm |
£77.65
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
Description
- Non-contaminating
- Impervious to chemical diffusion at high pressure
- Wide operating temperature range, -180 to +300°C
- Outstanding electrical insulation properties
- Individually tested
Specifications
Probe, PTFE Platinum Resistance | |
PTFE with Stainless Steel Core | |
Measuring temperature | |
50%, 6 to 14 sec. |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PRT100 Class A. TCR = 3850 ppm/degree celsius | |
PTFE Probe Holders and Compression Fittings | |
-180°C to +280°C |