missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ PTFE Polygonal Magnetic Stirring Bar

Ensure long-lasting durability and prevent demagnetization of Vitlab™ PFTE Polygonal Magnetic Stirring Bars via proper storage and not dropping them on hard surfaces (e.g. steel).

Brand:  VITLAB™ 301997

29.45 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10375711

  • £34.45 / Pack of 10
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Permanent magnet AlNiCo V core
  • Angled shape gives rise to significant turbulence
  • Achieves effective mixing, even at low spin rates
  • FTE
Specifications

Specifications

For preparation, separation and concentration
6 mm
PTFE
Polygon
Permanent magnet AlNiCo V core
25 mm
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers