Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Ensure long-lasting durability and prevent demagnetization of Vitlab™ PFTE Polygonal Magnetic Stirring Bars via proper storage and not dropping them on hard surfaces (e.g. steel).
Brand:  VITLAB™ 301797
 View more versions of this product
Product Code. 10674602
  • £31.00 / Pack of 10
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent magnet AlNiCo V core
  • Angled shape gives rise to significant turbulence
  • Achieves effective mixing, even at low spin rates
  • FTE
Specifications
Specifications
For preparation, separation and concentration
6 mm
PTFE
Polygon
Permanent magnet AlNiCo V core
15 mm
10/Pk.